Anti-MAPK4 Mouse Monoclonal Antibody [clone: OTI2D7]

Toimittaja: OriGene
TA505808
ORIGTA505808EA 621 EUR
ORIGTA505808
Anti-MAPK4 Mouse Monoclonal Antibody [clone: OTI2D7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK4 (mitogen-activated protein kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR