Anti-TLE2 Mouse Monoclonal Antibody [clone: OTI5A6]

Toimittaja: OriGene
TA504240
ORIGTA504240EA 621 EUR
ORIGTA504240
Anti-TLE2 Mouse Monoclonal Antibody [clone: OTI5A6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TLE2 (transducin-like enhancer of split 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR