Anti-NABP2 Mouse Monoclonal Antibody [clone: OTI1E11]

Toimittaja: OriGene
TA504022
ORIGTA504022EA 621 EUR
ORIGTA504022
Anti-NABP2 Mouse Monoclonal Antibody [clone: OTI1E11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NABP2 (nucleic acid binding protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR