Anti-PPARA Mouse Monoclonal Antibody [clone: OTI1E8]

Toimittaja: OriGene
TA503540
ORIGTA503540EA 621 EUR
ORIGTA503540
Anti-PPARA Mouse Monoclonal Antibody [clone: OTI1E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR