Anti-ZIM2 Mouse Monoclonal Antibody [clone: OTI5E12]

Toimittaja: OriGene
TA501906
ORIGTA501906EA 621 EUR
ORIGTA501906
Anti-ZIM2 Mouse Monoclonal Antibody [clone: OTI5E12]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZIM2 (zinc finger, imprinted 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR