Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9H8]

Toimittaja: OriGene
TA500080
ORIGTA500080EA 621 EUR
ORIGTA500080
Anti-AKT3 Mouse Monoclonal Antibody [clone: OTI9H8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1;50; Immunofluorescence: 1:100

Type: Primary
Antigen: AKT3 (v-akt murine thymoma viral oncogene homolog 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR