Anti-ALPPL2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA329491
ORIGTA329491EA 749 EUR
ORIGTA329491
Anti-ALPPL2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ALPPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR