Anti-Cxcl2 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA328580
ORIGTA328580EA 436 EUR
ORIGTA328580
Anti-Cxcl2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Cxcl2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR