Anti-CNTN1 Goat Polyclonal Antibody

Toimittaja: OriGene
TA326749
ORIGTA326749EA 785 EUR
ORIGTA326749
Anti-CNTN1 Goat Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: CNTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, , Mouse, Mouse, Pig, Rat,
Order Now

TEKNISET TIEDOT

Antigeenin symboli CNTN1
Environmentally Preferable
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Unconjugated
Reaktiivisuus Pig,Human,Dog,Rat,Mouse
Isäntä Goat
Isotyyppi IgG
Western blot Yes
Epitooppi internal
Immunogeeni Peptide with sequence C-QVTSQEYSARLEN, from the internal region of the protein sequence according to NP_001834.2; NP_778203.1.
Aminohappoluku 585 to 870
Sekvenssi C-QVTSQEYSARLEN
Lomake Liquid
Puhdistus purified by ammonium sulphate precipitation followed by antigen affinity chromatography
Ristiadsorptio No
Säilytyspuskuri Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA. Aliquot and store at -20°C. Minimize freezing and thawing.
Säilytyslämpötila –20 °C
Kuljetuslämpötila –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR