Anti-PHC3 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA326692
ORIGTA326692EA 792 EUR
ORIGTA326692
Anti-PHC3 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: PHC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR