Anti-MAP3K7 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA326164
ORIGTA326164EA 528 EUR
ORIGTA326164
Anti-MAP3K7 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:1000

Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR