Anti-ANAPC1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA315737
ORIGTA315737EA 528 EUR
ORIGTA315737
Anti-ANAPC1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: ANAPC1 (anaphase promoting complex subunit 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR