Anti-GPRC5C Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA315612
ORIGTA315612EA 528 EUR
ORIGTA315612
Anti-GPRC5C Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: GPRC5C (G protein-coupled receptor, class C, group 5, member C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR