Anti-SERGEF Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA315324
ORIGTA315324EA 528 EUR
ORIGTA315324
Anti-SERGEF Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: SERGEF (secretion regulating guanine nucleotide exchange factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR