Anti-CACNB2 Mouse Monoclonal Antibody [clone: S8b-1]

Toimittaja: OriGene
TA309311
ORIGTA309311EA 805 EUR
ORIGTA309311
Anti-CACNB2 Mouse Monoclonal Antibody [clone: S8b-1]
Vasta-aineet
Recommended Dilutions:

Type: Primary
Antigen: CACNB2 (calcium channel, voltage-dependent, beta 2 subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR