Anti-QSOX1 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA308282
ORIGTA308282EA 868 EUR
ORIGTA308282
Anti-QSOX1 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: QSOX1 (quiescin Q6 sulfhydryl oxidase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR