Anti-THEM4 Rabbit Polyclonal Antibody

Toimittaja: OriGene
TA306801
ORIGTA306801EA 792 EUR
ORIGTA306801
Anti-THEM4 Rabbit Polyclonal Antibody
Vasta-aineet
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: THEM4 (thioesterase superfamily member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR