Anti-CD33 Mouse Monoclonal Antibody [clone: 1C4]

Toimittaja: OriGene
CF506349
ORIGCF506349EA 833 EUR
ORIGCF506349
Anti-CD33 Mouse Monoclonal Antibody [clone: 1C4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR