Anti-MICAL1 Mouse Monoclonal Antibody [clone: 6C7]

Toimittaja: OriGene
CF501849
ORIGCF501849EA 833 EUR
ORIGCF501849
Anti-MICAL1 Mouse Monoclonal Antibody [clone: 6C7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MICAL1
Clonality: Monoclonal
Clone: 6C7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR