Anti-XRCC1 Mouse Monoclonal Antibody [clone: 2G8]

Toimittaja: OriGene
CF500879
ORIGCF500879EA 833 EUR
ORIGCF500879
Anti-XRCC1 Mouse Monoclonal Antibody [clone: 2G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: XRCC1
Clonality: Monoclonal
Clone: 2G8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR