Omnifit® EZ Glass Chromatography Columns for FPLC and LPLC
Toimittaja: OMNIFIT
006EZ-10-33-AA 006EZ-10-33-AF 006EZ-15-33-AA 006EZ-15-33-AF 006EZ-25-33-AA 006EZ-10-25-AF 006EZ-06-15-AA 006EZ-06-25-AA 006EZ-15-50-AF 006EZ-06-05-AA 006EZ-06-05-AF 006EZ-06-05-FF 006EZ-06-10-AA 006EZ-06-10-AF 006EZ-06-10-FF 006EZ-06-15-AF 006EZ-06-15-FF 006EZ-06-25-AF 006EZ-06-25-FF 006EZ-06-40-AA 006EZ-06-40-AF 006EZ-06-40-FF 006EZ-10-10-AA 006EZ-10-10-AF 006EZ-10-10-FF 006EZ-10-15-AA 006EZ-10-15-AF 006EZ-10-15-FF 006EZ-10-25-AA 006EZ-10-25-FF 006EZ-10-40-AA 006EZ-10-40-AF 006EZ-10-40-FF 006EZ-10-50-AA 006EZ-10-50-AF 006EZ-10-50-FF 006EZ-15-10-AA 006EZ-15-10-AF 006EZ-15-10-FF 006EZ-15-15-AA 006EZ-15-15-AF 006EZ-15-15-FF 006EZ-15-25-AA 006EZ-15-25-AF 006EZ-15-25-FF 006EZ-15-40-AA 006EZ-15-40-AF 006EZ-15-40-FF 006EZ-15-50-AA 006EZ-15-50-FF 006EZ-25-10-AA 006EZ-25-10-AF 006EZ-25-10-FF 006EZ-25-15-AA 006EZ-25-15-AF 006EZ-25-15-FF 006EZ-25-25-AA 006EZ-25-25-FF 006EZ-25-40-AA 006EZ-25-40-AF 006EZ-25-40-FF 006EZ-25-50-AA 006EZ-25-50-AF 006EZ-25-50-FF 006EZ-35-15-AF 006EZ-35-15-FF 006EZ-35-25-AA 006EZ-35-25-AF 006EZ-35-40-AA 006EZ-35-40-AF 006EZ-35-40-FF 006EZ-50-25-FF 006EZ-50-25-AA 006EZ-50-25-AF 006EZ-50-40-AA 006EZ-50-40-AF 006EZ-50-40-FF 006EZ-50-50-AA 006EZ-50-50-AF 006EZ-50-50-FF 006EZ-05-25-AA 006EZ-50-33AA 006EZ-05-40-AA 006EZ-05-33-AA
OMNI006EZ-10-33-AAEA 1720 EUR
OMNI006EZ-10-33-AA OMNI006EZ-10-33-AF OMNI006EZ-15-33-AA OMNI006EZ-15-33-AF OMNI006EZ-25-33-AA OMNI006EZ-10-25-AF OMNI006EZ-06-15-AA OMNI006EZ-06-25-AA OMNI006EZ-15-50-AF OMNI006EZ-06-05-AA OMNI006EZ-06-05-AF OMNI006EZ-06-05-FF OMNI006EZ-06-10-AA OMNI006EZ-06-10-AF OMNI006EZ-06-10-FF OMNI006EZ-06-15-AF OMNI006EZ-06-15-FF OMNI006EZ-06-25-AF OMNI006EZ-06-25-FF OMNI006EZ-06-40-AA OMNI006EZ-06-40-AF OMNI006EZ-06-40-FF OMNI006EZ-10-10-AA OMNI006EZ-10-10-AF OMNI006EZ-10-10-FF OMNI006EZ-10-15-AA OMNI006EZ-10-15-AF OMNI006EZ-10-15-FF OMNI006EZ-10-25-AA OMNI006EZ-10-25-FF OMNI006EZ-10-40-AA OMNI006EZ-10-40-AF OMNI006EZ-10-40-FF OMNI006EZ-10-50-AA OMNI006EZ-10-50-AF OMNI006EZ-10-50-FF OMNI006EZ-15-10-AA OMNI006EZ-15-10-AF OMNI006EZ-15-10-FF OMNI006EZ-15-15-AA OMNI006EZ-15-15-AF OMNI006EZ-15-15-FF OMNI006EZ-15-25-AA OMNI006EZ-15-25-AF OMNI006EZ-15-25-FF OMNI006EZ-15-40-AA OMNI006EZ-15-40-AF OMNI006EZ-15-40-FF OMNI006EZ-15-50-AA OMNI006EZ-15-50-FF OMNI006EZ-25-10-AA OMNI006EZ-25-10-AF OMNI006EZ-25-10-FF OMNI006EZ-25-15-AA OMNI006EZ-25-15-AF OMNI006EZ-25-15-FF OMNI006EZ-25-25-AA OMNI006EZ-25-25-FF OMNI006EZ-25-40-AA OMNI006EZ-25-40-AF OMNI006EZ-25-40-FF OMNI006EZ-25-50-AA OMNI006EZ-25-50-AF OMNI006EZ-25-50-FF OMNI006EZ-35-15-AF OMNI006EZ-35-15-FF OMNI006EZ-35-25-AA OMNI006EZ-35-25-AF OMNI006EZ-35-40-AA OMNI006EZ-35-40-AF OMNI006EZ-35-40-FF OMNI006EZ-50-25-FF OMNI006EZ-50-25-AA OMNI006EZ-50-25-AF OMNI006EZ-50-40-AA OMNI006EZ-50-40-AF OMNI006EZ-50-40-FF OMNI006EZ-50-50-AA OMNI006EZ-50-50-AF OMNI006EZ-50-50-FF OMNI006EZ-05-25-AA OMNI006EZ-50-33AA OMNI006EZ-05-40-AA OMNI006EZ-05-33-AA
Omnifit® EZ Glass Chromatography Columns for FPLC and LPLC
Kromatografiakolonnit
EZ columns are designed to suit the majority of standard laboratory chromatography applications. They are ideal for aqueous systems and compatible with solvents used in common liquid chromatography applications such as protein purification.
  • Available in seven diameters and a variety of lengths
  • Compatible with a variety of bulk media
  • Operating temperature: 4 to 40 °C
  • Glass column is made up of borosilicate glass
Economical modular columns for low pressure or gravity feed, providing even distribution of eluent to the medium bed, with or without flowMore Product Information