Multifunctional column mounted washdown bench scales, for standard industrial applications, Defender 5000
Toimittaja: OHAUS
30444793 30444791 30444792 30444794 30444795 30444796 30444797 30444798 30444799 30444800 30444801 30444802 30444803 30461436 30461437 30461568 30461569 30461570 30461571 30461572 30461573 30461574 30461575 30461576 30461577 30461578 30471130 30471131 30471132 30471133 30471134 30471135 30471136 30471137 30471158 30471159 30471160 30471161 30471162 30471163 30471164 30471165
OHAU30444793EA 1710 EUR
OHAU30444793 OHAU30444791 OHAU30444792 OHAU30444794 OHAU30444795 OHAU30444796 OHAU30444797 OHAU30444798 OHAU30444799 OHAU30444800 OHAU30444801 OHAU30444802 OHAU30444803 OHAU30461436 OHAU30461437 OHAU30461568 OHAU30461569 OHAU30461570 OHAU30461571 OHAU30461572 OHAU30461573 OHAU30461574 OHAU30461575 OHAU30461576 OHAU30461577 OHAU30461578 OHAU30471130 OHAU30471131 OHAU30471132 OHAU30471133 OHAU30471134 OHAU30471135 OHAU30471136 OHAU30471137 OHAU30471158 OHAU30471159 OHAU30471160 OHAU30471161 OHAU30471162 OHAU30471163 OHAU30471164 OHAU30471165
Multifunctional column mounted washdown bench scales, for standard industrial applications, Defender 5000
Vaa'at
The Defender 5000 series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, stainless steel frame and a stainless steel (single range models) or aluminium load cell (dual range models).
  • Large backlit display and alpha-numeric keypad with function keys
  • Indicator guides user with on-screen prompts
  • Multiple application modes and high storage capacity for greater flexibility
  • Multiple