Multifunctional front mounted washdown bench scales, Defender 5000
Toimittaja: OHAUS
30461565 30461566 30461431 30461432 30461433 30461434 30461435 30471124 30471125 30471126 30471127 30471128 30471129 30444765 30444766 30444767 30444788 30444789 30444790 30461428 30471152 30471153 30471154 30471155 30471156 30471157
OHAU30461565EA 1720 EUR
OHAU30461565 OHAU30461566 OHAU30461431 OHAU30461432 OHAU30461433 OHAU30461434 OHAU30461435 OHAU30471124 OHAU30471125 OHAU30471126 OHAU30471127 OHAU30471128 OHAU30471129 OHAU30444765 OHAU30444766 OHAU30444767 OHAU30444788 OHAU30444789 OHAU30444790 OHAU30461428 OHAU30471152 OHAU30471153 OHAU30471154 OHAU30471155 OHAU30471156 OHAU30471157
Multifunctional front mounted washdown bench scales, Defender 5000
Vaa'at
The Defender 5000 series multifunctional bench scales are ideal for numerous applications including production, packaging, inventory and shipping. Durably constructed to withstand harsh environments and equipped with a stainless steel platform, stainless steel frame and a stainless steel (single range models) or aluminium load cell (dual range models). The front mounted D52XW indicator is 304 stainless steel with sand blasted surface treatment, which offers protection to IP 68.
  • Large backlit display and alpha-numeric keypad with function keys
  • Indicator