Trays for digital/analogue rocking and waving shakers
Toimittaja: OHAUS
30400137 30400138 30400139
OHAU30400137EA 151 EUR
OHAU30400137 OHAU30400138 OHAU30400139
Trays for digital/analogue rocking and waving shakers
Haihdutusmaljat/Tarjottimet/Altaat Instrumenttialustat/-astiat
These trays expand the capacity of the shakers by adding a second tier to the shaker.
  • Easily attaches to the unit, includes tray to add a second tier for higher capacity applications
  • Second tier tray mounts 8,9 cm above the lower tray
Toimitustiedot: Tray includes hardware and a rubber mat.