Digital block heaters, Advanced
Toimittaja: OHAUS
30392082 30392061 30392089 30392096
OHAU30392082EA 1010 EUR
OHAU30392082 OHAU30392061 OHAU30392089 OHAU30392096
Digital block heaters, Advanced
Kuivahauteet Digitaaliset kuivahauteet
Designed for applications that require repeatable results and superior temperature stability. These multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures.
  • PID temperature control, with optional external RTD probe, offers a temperature stability as low as ±0,1 °C with a temperature uniformity as low as ±0,1 °C
  • Hot warning symbol light is illuminated when the temperature is above 40 °C
  • Overshoot protection ensures unit will