Block heaters, analogue
Toimittaja: OHAUS
30392047 30392054 30392068 30392075
OHAU30392047EA 527 EUR
OHAU30392047 OHAU30392054 OHAU30392068 OHAU30392075
Block heaters, analogue
Kuivahauteet Analogiset kuivahauteet
These high wattage, constant-temperature dry block heaters are economical, versatile and compact. The multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
  • PID temperature controller maintains precise temperature control
  • Samples are heated to temperature quickly and accurately
  • Has the temperature uniformity of ±0,1 °C at 37 °C within the block
  • Can be used for coagulation and RH studies
Each model accepts separate interchangeableMore Product Information