Cell factories, Nunc™ high density
Toimittaja: Thermo Fisher Scientific
169102 169118 169160
NUNC169102EA 1162 EUR
NUNC169102 NUNC169118 NUNC169160
Cell factories, Nunc™ high density
Bioreaktorit
Thermo Scientific™ Nunc™ High Density Cell Factory™ system offers greater surface area and yield than the standard Cell Factory™ system, or similar multi-tray systems for adherent cell culture. It offers an efficient, economic option for increasing production capacity and cell culture yield in the same footprint.
  • Certified Nunclon™∆ surface treatment ensures excellent conditions for cell attachment and growth
  • Increased yield resulting from 30% more surface area within the standard Cell Factory™ footprint
  • Increased labour and handling efficiencies by