Soluviljelylevyt, monikuoppaiset, poly-D-lysiini -päällystetty, Nunc™
Toimittaja: Thermo Fisher Scientific
152035 152025
735-0227EA 660 EUR
735-0227 NUNC152025
Soluviljelylevyt, monikuoppaiset, poly-D-lysiini -päällystetty, Nunc™
Kuoppalevyt Soluviljelylevyt, monikuoppaiset
PS, with lid. Useful in all areas of cell culture.
  • Raised well rims to lower risk of contamination
  • Excellent optical quality
  • Nunclon™Δ certified surface treatment
Ideal for culturing finicky cells that do not attach to the regular cell culture-treated surfaces.
Tilaustiedot: Dishes come packed in sleeves of five.