Anti-TRPV1 Rabbit Polyclonal Antibody

Toimittaja: Novus Biologicals
NBP1-71774 NBP1-71774SS
NOVUNBP1-71774EA 623 EUR
NOVUNBP1-71774 NOVUNBP1-71774SS
Anti-TRPV1 Rabbit Polyclonal Antibody
Vasta-aineet
The Vanilloid R1 / TRPV1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Vanilloid R1 / TRPV1. This antibody reacts with human, mouse. The Vanilloid R1 / TRPV1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR