Anti-C12orf5 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Toimittaja: Novus Biologicals
NBP1-49534F
NOVUNBP1-49534FEA 668 EUR
NOVUNBP1-49534F
Anti-C12orf5 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR