Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-6E10]

Toimittaja: Novus Biologicals
NBP1-18981 NBP1-18981SS
NOVUNBP1-18981EA 554 EUR
NOVUNBP1-18981 NOVUNBP1-18981SS
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-6E10]
Vasta-aineet
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-6E10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR