Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 6A10]

Toimittaja: Novus Biologicals
NB600-443 NB600-443SS
NOVUNB600-443EA 606 EUR
NOVUNB600-443 NOVUNB600-443SS
Anti-Aflatoxin B1 Mouse Monoclonal Antibody [clone: 6A10]
Vasta-aineet
Type: Primary
Antigen: Aflatoxin B1 (aflatoxin B1)
Clonality: monoclonal
Clone: 6A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR