Anti-LCAT Rabbit Polyclonal Antibody

Toimittaja: Novus Biologicals
NB400-107 NB400-107SS
NOVUNB400-107EA 591 EUR
NOVUNB400-107 NOVUNB400-107SS
Anti-LCAT Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: LCAT (lecithin-cholesterol acyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR