Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)

Toimittaja: Novus Biologicals
NB110-58749G
NOVUNB110-58749GEA 606 EUR
NOVUNB110-58749G
Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)
Vasta-aineet
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR