Anti-FOXP3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Toimittaja: Novus Biologicals
NB100-39002PE
NOVUNB100-39002PEEA 806 EUR
NOVUNB100-39002PE
Anti-FOXP3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR