RABS/isolator mittens, BioClean™ Clean Nitrile CGL
Toimittaja: Nitritex
CGL30NITM9_U CGL30NITM9
112-6121EA 445 EUR
112-6121 NITRCGL30NITM9
RABS/isolator mittens, BioClean™ Clean Nitrile CGL
BioClean RABS and isolator mittens are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
  • Nitrile, contains no natural latex proteins
  • 100% inspected and air leak tested
  • Suitable for autoclaving
  • Resistant to VHP, IPA and disinfectants
  • Ultra-clean surface ensures product protection
  • Specially designed to minimise hand fatigue
Designed for use in injectable manufacturing and pharmaceutical production areas, the CGL mittens are cleanroom processed and packed.
Suitable for use in Class 10, ISO 4 and EU GMPMore Product Information