Steam sterilisers, KS series
Toimittaja: NICKEL ELECTRO
KS1 KS2
NICKKS1EA 7550 EUR
NICKKS1 NICKKS2
Steam sterilisers, KS series
Sterilisaattorit Autoklaavit (höyrysterilointilaitteet)
This steriliser is a laboratory steriliser, using steam to sterilise small unwrapped instruments and non-porous equipment which displaces air passively from the chamber and its load by steam generated within the steriliser.
  • Double skinned walled chamber for insulation purposes
  • Removable shelves for ease of loading
  • Provides constant boiling water producing steam
The lid is designed to ensure condensed steam runs back into the chamber and can be removed using insulated lifting handles. These handles on both the unit and lid features an 18 mm diameterMore Product Information