PIG® leak diverter quick-deploy kit
Toimittaja: NEW PIG
TLS461-YW TLS903
NEWPTLS461-YWEA 325 EUR
NEWPTLS461-YW NEWPTLS903
PIG® leak diverter quick-deploy kit
Imeytysaineet ja imeytysliinat
Quick deploying leak diverter springs open fast to catch roof leaks. Simply pop it open.
  • Superfast to control leaks and protect people, inventory, and equipment
  • Hangs easily in problem spots to catch nuisance leaks and channel them to a floor drain or collection container
  • Nonpermeable tarp is vinyl coated for strength and durability; resists punctures, UV and mildew for long life
  • Pinched corners help funnel liquid toward center for better drainage
  • Connect flexible PVC drain hose (included) to channel liquid to floor drain or collection container