Low-profile leak diverters for pipes, PIG®
Toimittaja: NEW PIG
TLS190
NEWPTLS190EA 108 EUR
NEWPTLS190
Low-profile leak diverters for pipes, PIG®
Imeytysaineet ja imeytysliinat
Low-profile design is great for capturing leaks where full-size designs could not be used.
  • Low-profile design and compact size let target a specific leak in tight spaces
  • Non-permeable material is vinyl-coated for strength and durability
  • Resists punctures and mildew for long life
  • UV resistant up to 12 months for long-term outdoor use without degradation
  • Easy to hang from an overhead pipe
Built-in grommets and rubber straps, a standard 3/4" hose connection to safely channel liquid to floor drain or collection container (hose sold separately). Flow rateMore Product Information