PIG® HAM-O® universal mat pads and rolls
Toimittaja: NEW PIG
MAT116 MAT267 MAT268 MAT269
NEWPMAT116EA 422 EUR
NEWPMAT116 NEWPMAT267 NEWPMAT268 NEWPMAT269
PIG® HAM-O® universal mat pads and rolls
Imeytysaineet ja imeytysliinat
"Hamouflage" pattern hides grime so mat always looks clean. Scuff-resistant top layer stands up to foot traffic in aisles and walkways.
  • Exclusive "hamouflage" top layer saves on product and disposal costs; absorbed grime is invisible, so you'll leave mat in place longer and save money
  • Fast-wicking "diaper action" transfers liquids from the top layer into the absorbent inner layers; spills disappear in seconds
  • Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated
  • Under the Ham-O top