PIG® HazMat Spill kit in a clip-close bag
Toimittaja: NEW PIG
KITE391
NEWPKITE391EA 48.5 EUR
NEWPKITE391
PIG® HazMat Spill kit in a clip-close bag
Imeytysaineet ja imeytysliinat
PIG absorbents in low cost, easy to close, hang and carry bag.
  • PIG Sock to stop spreading spills; PIG Mat absorbs quickly
  • PIG HazMat absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bag helps make clean up easier
  • Durable see-through bag allows you to quickly and easily inspect spill kit supplies
  • Heavy gauge polythene bag helps protect absorbents from moisture and damage during storage to ensure maximum product performance in use
  • Convenient clip