PIG® HazMat chemical absorbent dikes
Toimittaja: NEW PIG
HA1010
NEWPHA1010EA 234 EUR
NEWPHA1010
PIG® HazMat chemical absorbent dikes
Imeytysaineet ja imeytysliinat
Your biggest spills require the stopping power of these large-diameter hazmat dikes.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide
  • Chemical-resistant sock won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of larger chemical spills; for diking smaller chemical spills, consider our smaller diameter PIG HazMat Socks
  • PP skin resists chemicals and tearing; reduces dust and holds