Drum funnels
Toimittaja: NEW PIG
DRM672-BK DRM452-BK
NEWPDRM672-BKEA 176 EUR
NEWPDRM672-BK NEWPDRM452-BK
Drum funnels
Suppilot Suodatussuppilot Tynnyrisuppilot
Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
  • Allows access to ³/₄" bung for use of liquid-level gauge
  • Level inside surface allows easy draining of containers, parts or filters
  • Zigzag ridges prevent splashes and catch parts or tools
These large diameter funnels covers entire drum top to minimize drips and spills.