Absorbent booms, oil-only, Versatile PIG®
Toimittaja: NEW PIG
BOM304 BOM405 BOM406 BOM408 BOM409
NEWPBOM304EA 285 EUR
NEWPBOM304 NEWPBOM405 NEWPBOM406 NEWPBOM408 NEWPBOM409
Absorbent booms, oil-only, Versatile PIG®
Imeytysaineet ja imeytysliinat
Booms made from PP skin (UV resistant up to 12 months), PP filler and a tough outer mesh which is UV resistant for long-term outdoor use without degradation. The PP filler is chemical-resistant and hydrophobic for containing and absorbing oil-based spills. The booms float at water surface for easy retrieval, even when saturated.
  • Rope running along length of boom withstands up to 400-lb. loads, adding strength for deployment and retrieval
  • Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together for greater length
  • Bright