Cold traps, with olives
Toimittaja: NEUBERT VOLUME GLASSWAERE
0725-70-45 07257045505235
NEUB0725-70-45EA 0 EUR
NEUB0725-70-45 NEUB07257045505235
Cold traps, with olives
Kylmäloukut
This cooling traps are made from borosilicate glass 3.3, with sleeve on the lower part, two 10 mm glass poles and ground clamp made of PP.