Specialty bags, Whirl-Pak®
Toimittaja: NASCO FORT ATKINSON
B01350WA B01324WA B01319WA
NASCB01350WAEA 422 EUR
NASCB01350WA 300-0667 NASCB01319WA
Specialty bags, Whirl-Pak®
Pussit ja sulkijat Näytepussit
Blended, virgin polyethylene film provides exceptional clarity and superior strength HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • For use in your meat and poultry facilities
  • Continuous perimeter seal: Maximum protection against leaks
Specially designed products for meat and poultry sampling, Poultry rinse polyethylene bag is sealed at the top to maintain sterility, and has a perforation for easy opening. It will hold smallMore Product Information