Racks for Whirl-Pak® bags
Toimittaja: NASCO FORT ATKINSON
B00751WA B01014WA B01533WA
NASCB00751WAEA 146 EUR
NASCB00751WA NASCB01014WA NASCB01533WA
Racks for Whirl-Pak® bags
Koeputkitelineet Pussitelineet
Storage racks for Whirl-Pak® bags. Perfect for storage and transportation.
  • 10 and 12 gauge steel fused together
  • Powder coated
  • Autoclavable
Rack is finished with an electrostatically applied powder coating. The result is a strong, durable finish which will stand up to many laboratory materials and the rack can be autoclaved. Max dry temperature is 148 °C and wet temperature is 260 °C.