Metric non-phthalate PVC tubing, Nalgene™
Toimittaja: Thermo Fisher Scientific
8703-0407 8703-0508 8703-0406 8703-0609 8703-0610 8703-0710 8703-0811 8703-0812 8703-0913 8703-1014 8703-1216 8703-1520
NALG8703-0407EA 115 EUR
NALG8703-0407 NALG8703-0508 NALG8703-0406 NALG8703-0609 NALG8703-0610 NALG8703-0710 NALG8703-0811 NALG8703-0812 NALG8703-0913 NALG8703-1014 NALG8703-1216 NALG8703-1520
Metric non-phthalate PVC tubing, Nalgene™
Letku Siirtoputki
Establish a leak-tight fit with metric fittings. Nalgene lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø. and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
  • Offered in true metric sizes to fit your lab instrumentation and equipment
Nalgene™ Metric non-phthalate PVC tubing is manufactured from the highest-grade resinMore Product Information