Hanat, sulkuhanat, säiliöille, Nalgene®
Toimittaja: Thermo Fisher Scientific
6420-0500 6420-0750
NALG6420-0500EA 68.9 EUR
NALG6420-0500 NALG6420-0750
Hanat, sulkuhanat, säiliöille, Nalgene®
Hanat
LDPE, white.
  • Good chemical resistance
  • Godd temperature resistance
Ensure safe and efficient liquid dispensing from tanks by using Nalgene™ spigots for storage tanks. Spigots are for use with Naglene™ tanks up to 100 gal. with factory installed threaded boss.