Pullo, kapea suuaukko, pakkauspullo, kierrekorkilla, Nalgene®
Toimittaja: Thermo Fisher Scientific
322089-0001 322089-0002 322089-0008
NALG322089-0001EA 1220 EUR
NALG322089-0001 NALG322089-0002 NALG322089-0008
Pullo, kapea suuaukko, pakkauspullo, kierrekorkilla, Nalgene®
Pullot Muovipullot
Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.
  • Excellent impact and chemical resistance
  • Good temperature resistance
  • Food-grade
  • Include opaque amber polypropylene closures
  • Translucent amber bottles provide light protection with the ability to see liquid level
  • Leakproof when used with Nalgene closures
Guaranteed leakproof,More Product Information
Order Now
TEKNISET TIEDOT
Environmentally Preferable Low Manufacturing Impact
Ympäristösertifikaatti NAR Certificate of Retirement